Mesitylacetic acid 98%
Dodavatel: Apollo Scientific
APOSOR30409-25GEA
5000
CZK
APOSOR30409-25G
APOSOR30409-100G
Mesitylacetic acid 98%
Mesitylacetic acid
Vzorec:
C₁₁H₁₄O₂ MW: 178,23 g/mol Bod tání: 167…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014368 CAS číslo: 4408-60-0 EINECS: 224-556-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...