Tisk…

6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride 95+%

APOSOR30672-10GEA 0 CZK
APOSOR30672-10G APOSOR30672-5G APOSOR30672-1G
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride 95+%
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride
Vzorec: C₇H₁₀Cl₂N₂
MW: 193,0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09607967
CAS číslo: 147740-02-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR