4-Bromo-6-chloroquinoline
Dodavatel: Apollo Scientific
APOSOR307509-1GEA
11640
CZK
APOSOR307509-1G
4-Bromo-6-chloroquinoline
4-Bromo-6-chloroquinoline
Vzorec:
C₉H₅BrClN MW: 242,5000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11505112 CAS číslo: 1070879-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...