1-Methyl-1H-indazole-7-boronic acid pinacol ester
APOSOR308057-1GEA
0
CZK
APOSOR308057-1G
1-Methyl-1H-indazole-7-boronic acid pinacol ester
1-Methyl-1H-indazole-7-boronic acid pinacol ester
Vzorec:
C₁₄H₁₉BN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD15071446 CAS číslo: 1313738-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...