Tisk…

4-Bromo-6-methoxyquinoline

Dodavatel: Apollo Scientific

APOSOR309245-1GEA 3770 CZK
APOSOR309245-1G
4-Bromo-6-methoxyquinoline
4-Bromo-6-methoxyquinoline
Vzorec: C₁₀H₈BrNO
MW: 238,0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437074
CAS číslo: 42881-66-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR