3-Amino-3-(4-methylphenyl)propionic acid 97%
Dodavatel: Apollo Scientific
APOSOR309422-5GEA
2280
CZK
APOSOR309422-5G
3-Amino-3-(4-methylphenyl)propionic acid 97%
3-Amino-3-(4-methylphenyl)propionic acid
Vzorec:
C₁₀H₁₃NO₂ MW: 179,2157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863249 CAS číslo: 68208-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...