Tisk…

2-Amino-4-(4-bromophenyl)-5-methylthiazole

Dodavatel: Apollo Scientific

APOSOR30953-1GEA 6850 CZK
APOSOR30953-1G
2-Amino-4-(4-bromophenyl)-5-methylthiazole
2-Amino-4-(4-bromophenyl)-5-methylthiazole
Vzorec: C₁₀H₉BrN₂S
MW: 269,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00170268
CAS číslo: 65705-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR