Tisk…

4-Bromo-1-ethoxy-2-nitrobenzene

Dodavatel: Apollo Scientific

APOSOR310855-1GEA 2900 CZK
APOSOR310855-1G APOSOR310855-5G
4-Bromo-1-ethoxy-2-nitrobenzene
4-Bromo-1-ethoxy-2-nitrobenzene
Vzorec: C₈H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD12756413
CAS číslo: 383869-51-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR