Tisk…

1-Naphthyl boronic acid neopentyl glycol ester

Dodavatel: Apollo Scientific

APOSOR310989-5GEA 2900 CZK
APOSOR310989-5G APOSOR310989-25G
1-Naphthyl boronic acid neopentyl glycol ester
1-Naphthyl boronic acid neopentyl glycol ester
Vzorec: C₁₅H₁₇BO₂
MW: 240,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318167
CAS číslo: 22871-77-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR