Tisk…

(R)-(+)-Tetrahydro-2-furoic acid

Dodavatel: Apollo Scientific

APOSOR315017-25GEA 0 CZK
APOSOR315017-25G APOSOR315017-100G APOSOR315017-5G
(R)-(+)-Tetrahydro-2-furoic acid
(R)-(+)-Tetrahydro-2-furoic acid

Chiral intermediate for pharmaceuticals and agrochemicals.

Vzorec: C₅H₈O₃
MW: 116,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211271
CAS číslo: 87392-05-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR