Tisk…

Cyclohexyldiphenylphosphine

Dodavatel: Apollo Scientific

APOSOR315482-1GEA 1870 CZK
APOSOR315482-1G APOSOR315482-5G APOSOR315482-25G
Cyclohexyldiphenylphosphine
Cyclohexyldiphenylphosphine
Vzorec: C₁₈H₂₁P
MW: 268,33 g/mol
Bod tání: 53 °C
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00046360
CAS číslo: 6372-42-5
EINECS: 228-904-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR