Tisk…

Boc-D-valinol

Dodavatel: Apollo Scientific
Varování

APOSOR316019-25GEA 0 CZK
APOSOR316019-25G APOSOR316019-5G
Boc-D-valinol
Boc-D-valinol
Vzorec: C₁₀H₂₁NO₃
MW: 203,28 g/mol
Bod varu: 218 °C
Hustota: 0,995 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00235960
CAS číslo: 106391-87-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR