2-(4-Chlorophenyl)ethyl isothiocyanate
APOSOR322070-500MGEA
0
CZK
APOSOR322070-500MG
2-(4-Chlorophenyl)ethyl isothiocyanate
2-(4-Chlorophenyl)ethyl isothiocyanate
Vzorec:
C₉H₈ClNS MW: 197,69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041127 CAS číslo: 17608-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...