Tisk…

(R)-(+)-1-(p-Tolyl)ethylamine

Dodavatel: Apollo Scientific

APOSOR322115-1GEA 2490 CZK
APOSOR322115-1G
(R)-(+)-1-(p-Tolyl)ethylamine
(R)-(+)-1-(p-Tolyl)ethylamine
Vzorec: C₉H₁₃N
MW: 135,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145202
CAS číslo: 4187-38-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR