Tisk…

(R)-(+)-N,N-Dimethyl-1-phenylethanamine

Dodavatel: Apollo Scientific

APOSOR322173-1GEA 3910 CZK
APOSOR322173-1G
(R)-(+)-N,N-Dimethyl-1-phenylethanamine
(R)-(+)-N,N-Dimethyl-1-phenylethanamine
Vzorec: C₁₀H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD00008857
CAS číslo: 19342-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR