3-Formyl-6-nitrochromone
APOSOR322296-2GEA
0
CZK
APOSOR322296-2G
3-Formyl-6-nitrochromone
3-Formyl-6-nitrochromone
Vzorec:
C₁₀H₅NO₅ MW: 219,1504 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00192184 CAS číslo: 42059-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...