(Methoxymethyl)diphenylphosphine oxide
APOSOR322328-5GEA
0
CZK
APOSOR322328-5G
(Methoxymethyl)diphenylphosphine oxide
(Methoxymethyl)diphenylphosphine oxide
Vzorec:
C₁₄H₁₅O₂P MW: 246,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015451 CAS číslo: 4455-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...