N-Cbz-L-phenylalaninol
Dodavatel: Apollo Scientific
APOSOR322372-2GEA
4670
CZK
APOSOR322372-2G
N-Cbz-L-phenylalaninol
N-Cbz-L-phenylalaninol
Vzorec:
C₁₇H₁₉NO₃ MW: 285,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191138 CAS číslo: 6372-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...