Tisk…

2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyde

APOSOR322398-500MGEA 0 CZK
APOSOR322398-500MG
2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyde
2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyde
Vzorec: C₁₁H₁₀N₂O
MW: 186,2099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059904
CAS číslo: 1015845-89-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR