(S)-(+)-1-(4-Chlorophenyl)ethyl isothiocyanate
APOSOR322430-1GEA
0
CZK
APOSOR322430-1G
(S)-(+)-1-(4-Chlorophenyl)ethyl isothiocyanate
(S)-(+)-1-(4-Chlorophenyl)ethyl isothiocyanate
Vzorec:
C₉H₈ClNS Storage Temperature: Ambient |
MDL Number:
MFCD04039327 CAS číslo: 737000-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...