Tisk…

(R)-(-)-Tetrahydrofurfurylamine

Dodavatel: Apollo Scientific

APOSOR322445-500MGEA 5000 CZK
APOSOR322445-500MG
(R)-(-)-Tetrahydrofurfurylamine
(R)-(-)-Tetrahydrofurfurylamine
Vzorec: C₅H₁₁NO
MW: 101,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192476
CAS číslo: 7202-43-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR