(R)-(-)-1-(4-Chlorophenyl)ethyl isothiocyanate
APOSOR322453-1GEA
0
CZK
APOSOR322453-1G
(R)-(-)-1-(4-Chlorophenyl)ethyl isothiocyanate
(R)-(-)-1-(4-Chlorophenyl)ethyl isothiocyanate
Vzorec:
C₉H₈ClNS MW: 197,69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039326 CAS číslo: 737000-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...