Tisk…

2-Hydroxythiazole

Dodavatel: Apollo Scientific

Varování

APOSOR322501-100MGEA 5390 CZK
APOSOR322501-100MG
2-Hydroxythiazole
2-Hydroxythiazole
Vzorec: C₃H₃NOS
MW: 101,1270 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02094166
CAS číslo: 82358-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR