8-Amino-3-(4-chlorophenyl)imidazo[1,2-a]pyridine 95%
Dodavatel: Apollo Scientific
APOSOR32642-1GEA
16570
CZK
APOSOR32642-1G
8-Amino-3-(4-chlorophenyl)imidazo[1,2-a]pyridine 95%
8-Amino-3-(4-chlorophenyl)imidazo[1,2-a]pyridine
Vzorec:
C₁₃H₁₀ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD00975015 CAS číslo: 338415-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...