Tisk…

2-(4-Methoxyphenyl)isonicotinonitrile 95%

Dodavatel: Apollo Scientific

APOSOR32701-1GEA 16570 CZK
APOSOR32701-1G
2-(4-Methoxyphenyl)isonicotinonitrile 95%
2-(4-Methoxyphenyl)isonicotinonitrile
Vzorec: C₁₃H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD07366546
CAS číslo: 885949-66-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR