4-Amino-3-(4-bromophenyl)-1H-pyrazole 95%
Dodavatel: Apollo Scientific
APOSOR33574-1GEA
12630
CZK
APOSOR33574-1G
4-Amino-3-(4-bromophenyl)-1H-pyrazole 95%
4-Amino-3-(4-bromophenyl)-1H-pyrazole
Vzorec:
C₉H₈BrN₃ Storage Temperature: Ambient |
MDL Number:
MFCD10758068 CAS číslo: 91857-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...