(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
Dodavatel: Apollo Scientific
APOSOR340015-250MGEA
6620
CZK
APOSOR340015-250MG
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
Vzorec:
C₄₈H₃₀O₂ MW: 638,7500 g/mol Bod tání: 239…242 °C Storage Temperature: Freezer |
MDL Number:
MFCD09027270 CAS číslo: 361342-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...