5-Bromo-2-chlorobenzamide 95+%
Dodavatel: Apollo Scientific
APOSOR346687-25GEA
23740
CZK
APOSOR346687-25G
APOSOR346687-5G
APOSOR346687-1G
5-Bromo-2-chlorobenzamide 95+%
5-Bromo-2-chlorobenzamide
Vzorec:
C₇H₅BrClNO MW: 234,4777 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00482914 CAS číslo: 188774-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...