Tisk…

4-tert-Butylbenzophenone 97%

Dodavatel: Apollo Scientific

APOSOR3478-5GEA 3050 CZK
APOSOR3478-5G APOSOR3478-25G APOSOR3478-10G
4-tert-Butylbenzophenone 97%
4-tert-Butylbenzophenone
Vzorec: C₁₇H₁₈O
Bod varu: 168…170 °C
Bod tání: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00017255
CAS číslo: 22679-54-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR