Tisk…

6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one

Dodavatel: Apollo Scientific

APOSOR350036-1GEA 5080 CZK
APOSOR350036-1G
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one
Vzorec: C₆H₃BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08689590
CAS číslo: 21594-52-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR