Tisk…

(S)-2-Amino-2-cyclohexylacetic acid

Dodavatel: Apollo Scientific

APOSOR350213-100GEA 3280 CZK
APOSOR350213-100G
(S)-2-Amino-2-cyclohexylacetic acid
(S)-2-Amino-2-cyclohexylacetic acid
Vzorec: C₈H₁₅NO₂
MW: 157,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311679
CAS číslo: 14328-51-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR