Tisk…

5-Nitro-6-methyluracil

Dodavatel: Apollo Scientific

APOSOR350278-5GEA 1460 CZK
APOSOR350278-5G
5-Nitro-6-methyluracil
5-Nitro-6-methyluracil
Vzorec: C5H5N3O4
MW: 171,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047362
CAS číslo: 16632-21-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR