Tisk…

1-Naphthalenemethanol

Dodavatel: Apollo Scientific

APOSOR350290-100GEA 2700 CZK
APOSOR350290-100G
1-Naphthalenemethanol
1-Naphthalenemethanol
Vzorec: C₁₁H₁₀O
MW: 158,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004044
CAS číslo: 4780-79-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR