Tisk…

4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%

Dodavatel: Apollo Scientific

APOSOR360151-1GEA 4080 CZK
APOSOR360151-1G
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester
Vzorec: C₁₆H₂₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD08704684
CAS číslo: 859169-20-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR