Tisk…

4-Isobutoxyphenylboronic acid 95+%

APOSOR360219-25GEA 0 CZK
APOSOR360219-25G APOSOR360219-10G APOSOR360219-100G
4-Isobutoxyphenylboronic acid 95+%
4-Isobutoxyphenylboronic acid
Vzorec: C₁₀H₁₅BO₃
MW: 194,0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798083
CAS číslo: 153624-44-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR