6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 98%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR361336-1GEA
18850
CZK
APOSOR361336-1G
APOSOR361336-250MG
6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 98%
6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester
Vzorec:
C₁₆H₂₀BNO₄ MW: 301,1453 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11858368 CAS číslo: 1256359-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...