Tisk…

1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific

APOSOR361532-1GEA 2030 CZK
APOSOR361532-1G
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester 98%
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester
Vzorec: C₂₀H₂₂BNO₄S
MW: 383,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276778
CAS číslo: 870717-93-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR