4-Amino-3-iodobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR3730-1GEA
0
CZK
APOSOR3730-1G
4-Amino-3-iodobenzoic acid
4-Amino-3-iodobenzoic acid
Vzorec:
C₇H₆INO₂ MW: 263,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01660550 CAS číslo: 2122-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...