Tisk…

4-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific

Varování

APOSOR3793-1GEA 787 CZK
APOSOR3793-1G
4-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid 98%
4-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid
Vzorec: C₁₅H₁₆BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09027212
CAS číslo: 913835-38-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR