3-Ethynylthiophene 96%
APOSOR40642-5GEA
0
CZK
APOSOR40642-5G
3-Ethynylthiophene 96%
3-Ethynylthiophene
Vzorec:
C₆H₄S MW: 108,16 g/mol Bod varu: 152…153 °C Hustota: 1,098 g/cm³ (25 °C) Bod vzplanutí: 46,1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039973 CAS číslo: 67237-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...