3-(4-Cyanophenylcarbamoyl)phenylboronic acid 95%
|
Varování
|
APOSOR4115-1GEA
4500
CZK
APOSOR4115-1G
APOSOR4115-5G
3-(4-Cyanophenylcarbamoyl)phenylboronic acid 95%
3-(4-Cyanophenylcarbamoyl)phenylboronic acid
Vzorec:
C₁₄H₁₁BN₂O₃ MW: 266,0600 g/mol Bod tání: 232…238 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659886 CAS číslo: 850567-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...