Tisk…

4-Methylcyclohexene-1-ylboronic acid 98%

Varování

APOSOR4241-1GEA 13890 CZK
APOSOR4241-1G APOSOR4241-500MG APOSOR4241-250MG
4-Methylcyclohexene-1-ylboronic acid 98%
4-Methylcyclohexene-1-ylboronic acid
Vzorec: C₇H₁₃BO₂
MW: 139,99 g/mol
Bod tání: 162…166 °C
Storage Temperature: Freezer
MDL Number: MFCD06659857
CAS číslo: 850567-92-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR