2-Pyridineboronic acid dimethyl ester 95%
|
Varování
|
APOSOR4244-1GEA
9960
CZK
APOSOR4244-1G
2-Pyridineboronic acid dimethyl ester 95%
2-Pyridineboronic acid dimethyl ester
Vzorec:
C₇H₁₀BNO₂ MW: 150,97 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03412109 CAS číslo: 136805-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...