Tisk…

7-Chloro-4-quinolineboronic acid pinacol ester 90%

APOSOR4309-1GEA 0 CZK
APOSOR4309-1G
7-Chloro-4-quinolineboronic acid pinacol ester 90%
7-Chloro-4-quinolineboronic acid pinacol ester
Vzorec: C₁₅H₁₇BClNO₂
MW: 289,56 g/mol
Bod tání: 81…87 °C
Storage Temperature: Ambient
MDL Number: MFCD07783020
CAS číslo: 871125-83-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR