D-Cyclopropylalinine
Dodavatel: Apollo Scientific
APOSOR451082-5GEA
8140
CZK
APOSOR451082-5G
D-Cyclopropylalinine
D-Cyclopropylalinine
Vzorec:
C₆H₁₁NO₂ MW: 129,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02259520 CAS číslo: 121786-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...