4-(Dimethylamino)benzhydrazide
|
Varování
|
APOSOR460025-1GEA
953
CZK
APOSOR460025-1G
4-(Dimethylamino)benzhydrazide
4-(Dimethylamino)benzhydrazide
Vzorec:
C₉H₁₃N₃O MW: 179,22 g/mol Bod tání: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014761 CAS číslo: 19353-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...