Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Czech Republic
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 Kč
Order Entry
Home
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine 95%
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine 95%
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
274676-47-0
:
Formula:
C₉H₁₂N₂
MDL Number:
MFCD06739289
:
148.209 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR480241-1G
OR480241-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 5 660,00 Kč
5 660,00 Kč
-
+
Add to cart
1 * 1 g
5 660,00 Kč
-
+
Add to cart
APOSOR480241-5G
OR480241-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 21 560,00 Kč
21 560,00 Kč
-
+
Add to cart
1 * 5 g
21 560,00 Kč
-
+
Add to cart
APOSOR480241-10G
OR480241-10G
Bottle
10 g
Log in
for availability
Log in
for availability
1 * 10 g
1 * 10 g - 36 370,00 Kč
36 370,00 Kč
-
+
Add to cart
1 * 10 g
36 370,00 Kč
-
+
Add to cart