Tisk…
(1R,2R)-trans-2-Aminocyclohexanol hydrochloride 97%
Dodavatel: Apollo Scientific
APOSOR480768-5GEA 1910 CZK
APOSOR480768-5G
(1R,2R)-trans-2-Aminocyclohexanol hydrochloride 97%
(1R,2R)-trans-2-Aminocyclohexanol hydrochloride
Vzorec: C₆H₁₄ClNO
MW: 151,6345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09259962
CAS číslo: 13374-31-7