Tisk…

3-Bromo-5-methylanisole 97%

Dodavatel: Apollo Scientific

APOSOR50833-25GEA 3110 CZK
APOSOR50833-25G
3-Bromo-5-methylanisole 97%
3-Bromo-5-methylanisole
Vzorec: C₈H₉BrO
MW: 201,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061916
CAS číslo: 29578-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR