Methyl imidazo[1,2-a]pyridine-7-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR52106-1GEA
1250
CZK
APOSOR52106-1G
Methyl imidazo[1,2-a]pyridine-7-carboxylate
Methyl imidazo[1,2-a]pyridine-7-carboxylate
Vzorec:
C₉H₈N₂O₂ MW: 176,18 g/mol Bod tání: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD09910350 CAS číslo: 86718-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...